Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278541.1 | NP_001265470.1 | 8 | 889 | ||
NM_001872.4 | NP_001863.3 | 9 | 1000 | |||
XM_017020393.1 | XP_016875882.1 | 9 | 943 | |||
GenBank mRNA | AB011969.1 | - | 8 | 841 | ||
AK290829.1 | - | 9 | 948 | |||
BC007057.1 | - | 9 | 957 | 3934520 | ||
BT006936.1 | - | 9 | 933 | |||
M75106.1 | - | 9 | 952 |