Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031847.2 | NP_001027017.1 | 8, 9 | 1038 | ||
NM_001876.3 | NP_001867.2 | 8, 9 | 1038 | |||
XM_005273762.2 | XP_005273819.1 | 8, 9 | 1014 | |||
XM_005273763.1 | XP_005273820.1 | 8, 9 | 1014 | |||
XM_017017220.1 | XP_016872709.1 | 8, 9 | 1147 | |||
GenBank mRNA | AK025307.1 | - | 3, 4 | 324 | ||
AK172798.1 | - | 8, 9 | 1024 | |||
AK309464.1 | - | 8, 9 | 1014 | |||
BC000185.1 | - | 8, 9 | 913 | 3352642 | ||
BT009791.1 | - | 7, 8 | 868 | |||
L39211.1 | - | 8, 9 | 944 |