Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000573.3 | NP_000564.2 | 34 | 5719 | ||
NM_000651.4 | NP_000642.3 | 42 | 7069 | |||
XM_006711166.2 | XP_006711229.1 | 42 | 7045 | |||
XM_011509205.1 | XP_011507507.1 | 42 | 7045 | |||
GenBank mRNA | BF900429.1 | - | 1 | 52 | ||
X05309.1 | - | 25 | 4075 | |||
Y00816.1 | - | 5606 |