Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004075.4 | NP_004066.1 | 7 | 1941 | ||
XM_017018832.1 | XP_016874321.1 | 8 | 2076 | |||
GenBank mRNA | AK098615.1 | - | 2 | 344 | ||
AK125915.1 | - | 9 | 1604 | |||
AK290552.1 | - | 7 | 1241 | |||
BC017466.1 | - | 7 | 1364 | |||
BC030519.1 | - | 7 | 1614 | 5169081 | ||
D83702.1 | - | 7 | 1631 |