Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014617.3 | NP_055432.2 | 2 | 247 | ||
GenBank mRNA | BM698524.1 | - | 2 | 247 | ||
EF426311.1 | - | 2 | 239 | |||
KT001516.1 | - | 2 | 249 |