Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130042.1 | NP_001123514.1 | 5 | 818 | ||
NM_001130043.1 | NP_001123515.1 | 4 | 779 | |||
NM_001889.3 | NP_001880.2 | 4 | 779 | |||
XM_011540747.1 | XP_011539049.1 | 4 | 398 | |||
XM_017000367.1 | XP_016855856.1 | 5 | 453 | |||
GenBank mRNA | AB209714.1 | - | 5 | 450 | ||
AK128794.1 | - | 4 | 384 | |||
AK223150.1 | - | 4 | 411 | |||
AK223201.1 | - | 4 | 411 | |||
BC039578.1 | - | 4 | 401 | 6067125 | ||
BC070058.1 | - | 5 | 460 | 5267616 | ||
DA256305.1 | - | 4 | 463 | |||
L13278.1 | - | 3 | 323 |