Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004077.2 | NP_004068.2 | 9 | 1176 | ||
GenBank mRNA | AF047042.1 | - | 9 | 986 | ||
AF053631.1 | - | 9 | 1151 | |||
AK074956.1 | - | 9 | 1024 | |||
AK095856.1 | - | 9 | 1011 | |||
AK098177.1 | - | 2 | 1008 | |||
AK293263.1 | - | 7 | 878 | |||
AK296249.1 | - | 10 | 1156 | |||
AK307625.1 | - | 5 | 593 | |||
BC000105.1 | - | 7 | 816 | |||
BC010106.1 | - | 9 | 1104 | 3542491 | ||
BC072016.1 | - | 9 | 1019 | 6484456 | ||
BT007414.1 | - | 3 | 380 | |||
BX640838.1 | - | 10 | 1233 | |||
DQ403132.1 | - | 3 | 320 |