Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004077.2 | NP_004068.2 | 10 | 1354 | ||
GenBank mRNA | AF047042.1 | - | 10 | 1164 | ||
AF053631.1 | - | 10 | 1329 | |||
AK074956.1 | - | 10 | 1202 | |||
AK095856.1 | - | 10 | 1189 | |||
AK293263.1 | - | 8 | 1056 | |||
AK296249.1 | - | 11 | 1334 | |||
AK307625.1 | - | 6 | 771 | |||
BC000105.1 | - | 8 | 994 | |||
BC010106.1 | - | 10 | 1282 | 3542491 | ||
BC072016.1 | - | 10 | 1197 | 6484456 | ||
BT007414.1 | - | 4 | 558 | |||
BX640838.1 | - | 11 | 1411 | |||
DQ403132.1 | - | 4 | 498 |