Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004077.2 | NP_004068.2 | 10 | 1381 | ||
GenBank mRNA | AF047042.1 | - | 10 | 1191 | ||
AF053631.1 | - | 10 | 1356 | |||
AK074956.1 | - | 10 | 1229 | |||
AK095856.1 | - | 10 | 1216 | |||
AK293263.1 | - | 8 | 1083 | |||
AK296249.1 | - | 11 | 1361 | |||
AK307625.1 | - | 6 | 798 | |||
BC000105.1 | - | 8 | 1021 | |||
BC010106.1 | - | 10 | 1309 | 3542491 | ||
BC072016.1 | - | 10 | 1224 | 6484456 | ||
BT007414.1 | - | 4 | 585 | |||
BX640838.1 | - | 11 | 1438 | |||
DQ403132.1 | - | 4 | 525 |