Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CSF2RA SIRNA Assays ›
Gene Symbol:
CSF2RA
Gene Name:
colony stimulating factor 2 receptor alpha subunit
Gene Aliases:
CD116, CDw116, CSF2R, CSF2RAX, CSF2RAY, CSF2RX, CSF2RY, GM-CSF-R-alpha, GMCSFR, GMR, SMDP4
Chromosome Location:
Chr.X: 1268800 - 1325097 on Build GRCh38
UniGene ID:
Hs.520937
Species:
Human
siRNA ID
HSS102363
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CSF2RA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001161529.1
NP_001155001.1
8
921
NM_001161530.1
NP_001155002.1
5
625
NM_001161531.1
NP_001155003.1
7
793
NM_001161532.1
NP_001155004.1
5
562
NM_006140.4
NP_006131.2
7
793
NM_172245.2
NP_758448.1
7
769
NM_172246.2
NP_758449.1
7
793
NM_172247.2
NP_758450.1
5
625
NM_172249.2
NP_758452.1
7
793
NR_027760.1
7
793
XM_011546165.2
XP_011544467.1
6
671
XM_011546166.1
XP_011544468.1
6
671
XM_011546167.2
XP_011544469.1
6
671
XM_011546168.2
XP_011544470.1
6
671
XM_011546169.1
XP_011544471.1
6
671
XM_011546170.2
XP_011544472.1
6
671
XM_011546173.1
XP_011544475.1
6
671
XM_011546174.2
XP_011544476.1
6
671
XM_011546175.2
XP_011544477.1
6
671
XM_017029287.1
XP_016884776.1
7
783
XM_017029288.1
XP_016884777.1
7
760
GenBank mRNA
AK293086.1
-
7
751
AK301395.1
-
5
544
AK309630.1
-
7
754
AL833597.1
-
7
730
BC002635.1
-
7
769
3606186
BC071835.1
-
8
891
4650755
DQ841258.1
-
5
599
L29348.1
-
7
748
L29349.1
-
7
748
M64445.1
-
7
761
M73832.1
-
5
617
U93096.1
-
7
748
X17648.1
-
7
748
X54935.1
-
5
599
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
peptidyl-tyrosine phosphorylation
granulocyte colony-stimulating factor signaling pathway
positive regulation of GTPase activity
cellular protein metabolic process
cellular response to granulocyte macrophage colony-stimulating factor stimulus
protein tyrosine kinase activity
receptor activity
granulocyte colony-stimulating factor receptor activity
Ras guanyl-nucleotide exchange factor activity
protein binding
granulocyte colony-stimulating factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0