Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSNK1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001894.4 | NP_001885.1 | 4 | 394 | ||
NM_152221.2 | NP_689407.1 | 4 | 544 | |||
GenBank mRNA | AB024597.1 | - | 4 | 394 | ||
AB091043.1 | - | 4 | 360 | |||
AK223498.1 | - | 4 | 441 | |||
BC006490.2 | - | 4 | 349 | 3937782 | ||
BQ642398.1 | - | 528 | ||||
BT019831.1 | - | 3 | 283 | |||
CF594072.1 | - | 4 | 568 | |||
CR456429.1 | - | 3 | 296 | |||
L37043.1 | - | 3 | 305 |
Gene Symbol: LOC400927-CSNK1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289912.1 | NP_001276841.1 | 8 | 749 | ||
GenBank mRNA | AK092269.1 | - | 8 | 749 |