Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CSNK1G3 SIRNA Assays ›
Gene Symbol:
CSNK1G3
Gene Name:
casein kinase 1 gamma 3
Gene Aliases:
CKI-gamma 3, CSNK1G3L
Chromosome Location:
Chr.5: 123512099 - 123617045 on Build GRCh38
UniGene ID:
Hs.129206
Species:
Human
siRNA ID
HSS102392
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CSNK1G3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001031812.3
NP_001026982.1
5
1077
NM_001044723.2
NP_001038188.1
4
605
NM_001270572.1
NP_001257501.1
4
605
NM_001270573.1
NP_001257502.1
4
652
NM_001270574.1
NP_001257503.1
2
541
NM_004384.4
NP_004375.2
4
605
XM_005271891.1
XP_005271948.1
5
1077
XM_005271892.2
XP_005271949.1
5
1077
XM_005271894.2
XP_005271951.1
5
1077
XM_005271895.2
XP_005271952.1
5
1077
XM_005271896.1
XP_005271953.1
5
1077
XM_017009057.1
XP_016864546.1
6
1152
XM_017009058.1
XP_016864547.1
8
1361
XM_017009059.1
XP_016864548.1
7
1220
XM_017009060.1
XP_016864549.1
7
1276
XM_017009061.1
XP_016864550.1
6
1152
XM_017009062.1
XP_016864551.1
6
711
XM_017009063.1
XP_016864552.1
6
1152
XM_017009064.1
XP_016864553.1
6
1152
XM_017009065.1
XP_016864554.1
6
1152
XM_017009066.1
XP_016864555.1
7
1220
XM_017009067.1
XP_016864556.1
6
1152
XM_017009068.1
XP_016864557.1
6
1152
XM_017009069.1
XP_016864558.1
6
1145
XM_017009070.1
XP_016864559.1
5
1077
GenBank mRNA
AF049089.1
-
4
388
AF049090.1
-
4
388
AK289405.1
-
4
550
AK299739.1
-
4
491
AK316019.1
-
2
380
BC047567.1
-
5
729
4817058
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
cellular protein modification process
endocytosis
signal transduction
regulation of cell shape
Wnt signaling pathway
peptidyl-serine phosphorylation
protein kinase activity
protein serine/threonine kinase activity
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-x88h5:80/100.66.78.96:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline