Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004386.2 | NP_004377.2 | 8 | 2056 | ||
GenBank mRNA | AB210004.1 | - | 8 | 2000 | ||
AF026547.1 | - | 7 | 1958 | |||
AK297395.1 | - | 2 | 362 | |||
AK309138.1 | - | 2 | 528 |