Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300965.1 | NP_001287894.1 | 4 | 374 | ||
NM_001321.2 | NP_001312.1 | 4 | 399 | |||
GenBank mRNA | AB209321.1 | - | 1 | 4551 | ||
AY349166.1 | - | 4 | 354 | |||
BC000992.2 | - | 4 | 347 | 3448693 | ||
BT019913.1 | - | 3 | 295 | |||
BX461295.2 | - | 4 | 421 | |||
CN408330.1 | - | 4 | 345 | |||
U46006.1 | - | 3 | 295 | |||
U57646.1 | - | 4 | 351 |