Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001901.2 | NP_001892.1 | 3, 4 | 756 | ||
GenBank mRNA | AK092280.1 | - | 3, 4 | 607 | ||
AK125220.1 | - | 3, 4 | 541 | |||
AK290884.1 | - | 3, 4 | 750 | |||
AK315207.1 | - | 3, 4 | 750 | |||
AY395801.1 | - | 3, 4 | 673 | |||
AY550024.1 | - | 3, 4 | 663 | |||
BC087839.1 | - | 3, 4 | 732 | 6181580 | ||
BT019794.1 | - | 3, 4 | 550 | |||
BT019795.1 | - | 3, 4 | 550 | |||
CR541734.1 | - | 3, 4 | 550 | |||
CR541759.1 | - | 3, 4 | 550 | |||
M92934.1 | - | 3, 4 | 679 | |||
U14750.1 | - | 2, 3 | 330 | |||
X78947.1 | - | 3, 4 | 695 |