Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301237.1 | NP_001288166.1 | 1 | 173 | ||
NM_001905.3 | NP_001896.2 | 5 | 1009 | |||
NR_125440.1 | 5 | 1009 | ||||
GenBank mRNA | AK130549.1 | - | 4 | 595 | ||
AK225899.1 | - | 5 | 513 | |||
AK297386.1 | - | 5 | 605 | |||
AK303797.1 | - | 1 | 102 | |||
AK304817.1 | - | 5 | 544 | |||
BC009408.1 | - | 5 | 616 | 3355881 | ||
DA639348.1 | - | 1 | 173 | |||
X52142.1 | - | 5 | 540 |