Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001335.3 | NP_001326.2 | 10 | 1134 | ||
GenBank mRNA | AF013611.1 | - | 10 | 1092 | ||
BC035637.1 | - | 10 | 1097 | |||
BC048255.1 | - | 10 | 1134 | 5221520 | ||
BM996548.1 | - | 1 | 177 |