Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001335.3 | NP_001326.2 | 8, 9 | 855 | ||
GenBank mRNA | AF013611.1 | - | 8, 9 | 813 | ||
BC035637.1 | - | 8, 9 | 818 | |||
BC048255.1 | - | 8, 9 | 855 | 5221520 | ||
BM996548.1 | - | 2, 3 | 457 |