Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYLC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001340.4 | NP_001331.1 | 1, 2 | 89 | ||
GenBank mRNA | AK313349.1 | - | 1, 2 | 76 | ||
BC058038.1 | - | 1, 2 | 93 | |||
BC114547.1 | - | 1, 2 | 22 | 40005379 | ||
Z46788.1 | - | 1, 2 | 22 |