Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042355.1 | NP_001035814.1 | 18 | 3084 | ||
NM_001042412.1 | NP_001035877.1 | 18 | 3108 | |||
NM_015247.2 | NP_056062.1 | 20 | 3230 | |||
XM_005255812.2 | XP_005255869.1 | 19 | 3181 | |||
XM_006721149.1 | XP_006721212.1 | 19 | 3245 | |||
XM_011522907.2 | XP_011521209.1 | 18 | 3007 | |||
XM_011522908.1 | XP_011521210.1 | 18 | 3069 | |||
XM_017022977.1 | XP_016878466.1 | 18 | 3379 | |||
XM_017022978.1 | XP_016878467.1 | 18 | 3175 | |||
XM_017022979.1 | XP_016878468.1 | 18 | 3135 | |||
XM_017022980.1 | XP_016878469.1 | 18 | 3168 | |||
XM_017022981.1 | XP_016878470.1 | 18 | 3036 | |||
GenBank mRNA | AF161542.1 | - | 17 | 3057 | ||
AJ250014.1 | - | 20 | 3206 | |||
AK000187.1 | - | 13 | 1886 | |||
AK024348.1 | - | 16 | 2591 | |||
AK292975.1 | - | 18 | 3066 | |||
BC012342.1 | - | 18 | 3082 | 4552767 |