Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2C9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000771.3 | NP_000762.2 | 9 | 1432 | ||
XM_017015758.1 | XP_016871247.1 | 10 | 1852 | |||
GenBank mRNA | AK289420.1 | - | 9 | 1432 | ||
BC125054.1 | - | 9 | 1407 | 40116637 | ||
D00173.1 | - | 9 | 1398 | |||
M15331.1 | - | 9 | 1392 | |||
M21939.1 | - | 9 | 1393 | |||
M21940.1 | - | 7 | 1086 | |||
M61855.1 | - | 9 | 1419 | |||
M61857.1 | - | 9 | 1417 | |||
S46963.1 | - | 9 | 1370 |