Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2C18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000772.2 | NP_000763.1 | 1 | 454 | ||
NM_001128925.1 | NP_001122397.1 | 1 | 454 | |||
GenBank mRNA | AK223510.1 | - | 1 | 153 | ||
AK313403.1 | - | 1 | 153 | |||
BC069666.1 | - | 1 | 322 | 7262569 | ||
BC096257.1 | - | 1 | 173 | 40006072 | ||
BC096258.1 | - | 1 | 173 | 40006073 | ||
BC096260.1 | - | 1 | 173 | 40006076 | ||
BJ993698.1 | - | 1 | 454 | |||
M61853.1 | - | 1 | 170 | |||
M61856.1 | - | 1 | 327 |
Gene Symbol: CYP2C9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015758.1 | XP_016871247.1 | 1 | 159 |