Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000773.3 | NP_000764.1 | 4 | 623 | ||
GenBank mRNA | AF084225.1 | - | 4 | 497 | ||
AF182276.1 | - | 4 | 598 | |||
AJ853939.1 | - | 3 | 329 | |||
AJ853940.1 | - | 2 | 179 | |||
AK290822.1 | - | 4 | 623 | |||
BC067433.1 | - | 4 | 593 | 7002160 | ||
BC067435.1 | - | 4 | 594 | 7002164 | ||
DQ149222.1 | - | 4 | 607 | |||
J02625.1 | - | 4 | 602 |