Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000779.3 | NP_000770.2 | 8 | 1012 | ||
NM_001099772.1 | NP_001093242.1 | 8 | 1015 | |||
NM_001319161.1 | NP_001306090.1 | 7 | 970 | |||
NM_001319162.1 | NP_001306091.1 | 6 | 828 | |||
NM_001319163.1 | NP_001306092.1 | 6 | 825 | |||
NR_135003.1 | 8 | 942 | ||||
XM_011540832.1 | XP_011539134.1 | 8 | 1016 | |||
XM_011540833.1 | XP_011539135.1 | 7 | 648 | |||
XM_017000466.1 | XP_016855955.1 | 7 | 919 | |||
GenBank mRNA | AF491285.1 | - | 8 | 928 | ||
AK225576.1 | - | 8 | 964 | |||
AK225578.1 | - | 8 | 964 | |||
AK300921.1 | - | 6 | 825 | |||
AK300941.1 | - | 6 | 675 | |||
AK307715.1 | - | 8 | 1086 | |||
AK313170.1 | - | 8 | 967 | |||
AY064484.1 | - | 8 | 926 | |||
AY064485.1 | - | 8 | 928 | |||
AY064486.1 | - | 8 | 928 | |||
AY151048.1 | - | 8 | 931 | |||
AY151049.1 | - | 7 | 886 | |||
BC017758.1 | - | 8 | 967 | 4693225 | ||
CD013981.1 | - | 8 | 876 | |||
J02871.1 | - | 8 | 940 | |||
X16699.1 | - | 8 | 928 |