Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP11A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000781.2 | NP_000772.2 | 6 | 1166 | ||
NM_001099773.1 | NP_001093243.1 | 6 | 1240 | |||
GenBank mRNA | AK056794.1 | - | 6 | 1235 | ||
AK225715.1 | - | 5 | 754 | |||
AK292300.1 | - | 6 | 1072 | |||
AK292301.1 | - | 5 | 1030 | |||
AK293211.1 | - | 7 | 1032 | |||
AK300178.1 | - | 6 | 982 | |||
AY603498.1 | - | 2 | 54 | |||
BC032329.1 | - | 6 | 1055 | 5165263 | ||
KU178011.1 | - | 6 | 1011 | |||
M14565.1 | - | 6 | 1055 | |||
M28253.1 | - | 2 | 165 |