Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004080.2 | NP_004071.1 | 22 | 2193 | ||
NM_145695.2 | NP_663733.1 | 22 | 2193 | |||
XM_005249628.2 | XP_005249685.1 | 23 | 2721 | |||
XM_005249629.2 | XP_005249686.1 | 23 | 2718 | |||
XM_005249630.2 | XP_005249687.1 | 22 | 2234 | |||
XM_005249631.3 | XP_005249688.1 | 21 | 2449 | |||
XM_011515153.2 | XP_011513455.1 | 23 | 2559 | |||
XM_011515154.2 | XP_011513456.1 | 23 | 2275 | |||
XM_011515156.2 | XP_011513458.1 | 22 | 2231 | |||
XM_011515157.2 | XP_011513459.1 | 22 | 2238 | |||
XM_011515158.2 | XP_011513460.1 | 21 | 2374 | |||
XM_017011783.1 | XP_016867272.1 | 23 | 2612 | |||
XM_017011784.1 | XP_016867273.1 | 23 | 2272 | |||
XM_017011785.1 | XP_016867274.1 | 22 | 2506 | |||
XM_017011786.1 | XP_016867275.1 | 22 | 2486 | |||
XM_017011787.1 | XP_016867276.1 | 22 | 2236 | |||
XM_017011788.1 | XP_016867277.1 | 22 | 2522 | |||
XM_017011789.1 | XP_016867278.1 | 22 | 2239 | |||
XM_017011790.1 | XP_016867279.1 | 23 | 2558 | |||
GenBank mRNA | AB018261.1 | - | 19 | 1822 | ||
AK304719.1 | - | 21 | 2449 | |||
BC105005.1 | - | 22 | 2073 | 8144008 | ||
BC143632.1 | - | 22 | 2070 | 9052145 | ||
BC143633.1 | - | 21 | 2032 |