Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGKG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080744.1 | NP_001074213.1 | 13 | 1717 | ||
NM_001080745.1 | NP_001074214.1 | 12 | 1600 | |||
NM_001346.2 | NP_001337.2 | 13 | 1717 | |||
GenBank mRNA | AB209033.1 | - | 12 | 1443 | ||
AK304440.1 | - | 12 | 1598 | |||
AK314192.1 | - | 13 | 1662 | |||
AY737485.1 | - | 11 | 1295 | |||
BC089411.1 | - | 13 | 1471 | 3081161 | ||
BC112363.1 | - | 11 | 1323 | 40054027 | ||
D26135.1 | - | 13 | 1696 |