Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291963.1 | NP_001278892.1 | 1 | 227 | ||
NM_004394.2 | NP_004385.1 | 1 | 227 | |||
GenBank mRNA | AK298670.1 | - | 1 | 180 | ||
AK311783.1 | - | 1 | 106 | |||
BC002726.2 | - | 1 | 184 | 3631886 | ||
BI823002.1 | - | 1 | 58 | |||
BP265421.1 | - | 1 | 188 | |||
CR542184.1 | - | 1 | 20 | |||
DC395850.1 | - | 1 | 227 | |||
X76105.1 | - | 1 | 179 |