Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291963.1 | NP_001278892.1 | 2 | 337 | ||
NM_004394.2 | NP_004385.1 | 2 | 337 | |||
GenBank mRNA | AK298670.1 | - | 2 | 290 | ||
AK311783.1 | - | 2 | 216 | |||
BC002726.2 | - | 2 | 294 | 3631886 | ||
BI823002.1 | - | 2 | 168 | |||
BP265421.1 | - | 2 | 298 | |||
CR542184.1 | - | 2 | 130 | |||
X76105.1 | - | 2 | 289 |