Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DAPK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001348.2 | NP_001339.1 | 4 | 818 | ||
XM_005259508.1 | XP_005259565.1 | 4 | 693 | |||
GenBank mRNA | AB007144.1 | - | 3 | 614 | ||
AB022341.1 | - | 3 | 566 | |||
AB265224.1 | - | 1 | 31 | |||
AK027590.1 | - | 4 | 735 | |||
AK074799.1 | - | 3 | 590 | |||
AK097643.1 | - | 4 | 629 | |||
BC126430.1 | - | 3 | 557 | 8992146 | ||
BC126432.1 | - | 3 | 555 | 8992148 |