Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079862.2 | NP_001073331.1 | 2 | 166 | ||
NM_001079863.1 | NP_001073332.1 | 2 | 286 | |||
NM_001178017.1 | NP_001171488.1 | 2 | 305 | |||
NM_001178041.2 | NP_001171512.1 | 3 | 292 | |||
NM_001178042.2 | NP_001171513.1 | 3 | 247 | |||
NM_001178043.2 | NP_001171514.1 | 2 | 196 | |||
NM_001282633.1 | NP_001269562.1 | 3 | 282 | |||
NM_001282634.1 | NP_001269563.1 | 3 | 252 | |||
NM_001282635.1 | NP_001269564.1 | 3 | 282 | |||
NM_001282636.1 | NP_001269565.1 | 2 | 196 | |||
NM_020548.7 | NP_065438.1 | 3 | 277 | |||
NR_104221.1 | 2 | 445 | ||||
GenBank mRNA | AM000001.1 | - | 2 | 283 | ||
BC006466.1 | - | 2 | 81 | |||
BC062996.1 | - | 3 | 246 | 5168729 | ||
BM768076.1 | - | 2 | 113 | |||
BP294452.1 | - | 2 | 108 | |||
BU599304.1 | - | 2 | 169 | |||
CR456956.1 | - | 1 | 38 | |||
FM213122.1 | - | 2 | 143 | |||
FM213123.1 | - | 3 | 229 | |||
FM213124.1 | - | 3 | 224 | |||
FM213125.1 | - | 3 | 199 | |||
FM213126.1 | - | 3 | 194 | |||
FM213127.1 | - | 2 | 305 | |||
FM213128.1 | - | 3 | 239 | |||
FM213129.1 | - | 2 | 196 | |||
FM213130.1 | - | 2 | 445 | |||
FM213131.1 | - | 3 | 282 | |||
FM213132.1 | - | 3 | 174 | |||
M14200.1 | - | 2 | 108 | |||
M15887.1 | - | 2 | 95 |