Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079862.2 | NP_001073331.1 | 2 | 237 | ||
NM_001079863.1 | NP_001073332.1 | 2 | 357 | |||
NM_001178017.1 | NP_001171488.1 | 2 | 376 | |||
NM_001178041.2 | NP_001171512.1 | 3 | 363 | |||
NM_001178042.2 | NP_001171513.1 | 3 | 318 | |||
NM_001178043.2 | NP_001171514.1 | 2 | 267 | |||
NM_001282633.1 | NP_001269562.1 | 3 | 353 | |||
NM_001282634.1 | NP_001269563.1 | 3 | 323 | |||
NM_001282635.1 | NP_001269564.1 | 3 | 353 | |||
NM_001282636.1 | NP_001269565.1 | 2 | 267 | |||
NM_020548.7 | NP_065438.1 | 3 | 348 | |||
NR_104221.1 | 2 | 516 | ||||
GenBank mRNA | AM000001.1 | - | 2 | 354 | ||
BC006466.1 | - | 2 | 152 | |||
BC062996.1 | - | 3 | 317 | 5168729 | ||
BM768076.1 | - | 2 | 184 | |||
BP294452.1 | - | 2 | 179 | |||
BP318721.1 | - | 1 | 41 | |||
BU599304.1 | - | 2 | 240 | |||
CR456956.1 | - | 1 | 109 | |||
FM213122.1 | - | 2 | 214 | |||
FM213123.1 | - | 3 | 300 | |||
FM213124.1 | - | 3 | 295 | |||
FM213125.1 | - | 3 | 270 | |||
FM213126.1 | - | 3 | 265 | |||
FM213127.1 | - | 2 | 376 | |||
FM213128.1 | - | 3 | 310 | |||
FM213129.1 | - | 2 | 267 | |||
FM213130.1 | - | 2 | 516 | |||
FM213131.1 | - | 3 | 353 | |||
FM213132.1 | - | 3 | 245 | |||
M14200.1 | - | 2 | 179 | |||
M15887.1 | - | 2 | 166 |