Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135040.2 | NP_001128512.1 | 6 | 981 | ||
NM_001135041.2 | NP_001128513.1 | 4 | 704 | |||
NM_001190836.1 | NP_001177765.1 | 6 | 924 | |||
NM_001190837.1 | NP_001177766.1 | 8 | 1020 | |||
NM_004082.4 | NP_004073.2 | 9 | 1041 | |||
NM_023019.3 | NP_075408.1 | 4 | 704 | |||
NR_033935.1 | 6 | 924 | ||||
GenBank mRNA | AB209625.1 | - | 1 | 944 | ||
AK297286.1 | - | 6 | 676 | |||
AK314352.1 | - | 6 | 925 | |||
AK315014.1 | - | 4 | 496 | |||
BC071583.1 | - | 4 | 621 | 4817849 | ||
BX640799.1 | - | 9 | 820 | |||
BX647925.1 | - | 6 | 685 | |||
BX647972.1 | - | 6 | 704 | |||
X98801.1 | - | 7 | 678 |