Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DFNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127453.1 | NP_001120925.1 | 7 | 971 | ||
NM_001127454.1 | NP_001120926.1 | 6 | 986 | |||
NM_004403.2 | NP_004394.1 | 7 | 1216 | |||
XM_017011802.1 | XP_016867291.1 | 6 | 742 | |||
GenBank mRNA | AF007790.2 | - | 6 | 871 | ||
AF073308.1 | - | 7 | 940 | |||
AK094714.1 | - | 9 | 1544 | |||
AK290775.1 | - | 7 | 947 | |||
AK314402.1 | - | 7 | 956 | |||
BC099911.1 | - | 7 | 932 | 5287362 | ||
BC125065.1 | - | 7 | 938 | 40117291 | ||
BC125066.1 | - | 6 | 708 |