Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NQO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000903.2 | NP_000894.1 | 2 | 331 | ||
NM_001025433.1 | NP_001020604.1 | 2 | 331 | |||
NM_001025434.1 | NP_001020605.1 | 2 | 331 | |||
NM_001286137.1 | NP_001273066.1 | 2 | 369 | |||
GenBank mRNA | AK223050.1 | - | 2 | 243 | ||
AK297125.1 | - | 2 | 261 | |||
AK297979.1 | - | 2 | 261 | |||
AK298896.1 | - | 3 | 181 | |||
AK307533.1 | - | 2 | 193 | |||
AK312368.1 | - | 2 | 257 | |||
AK316246.1 | - | 2 | 154 | |||
BC007659.2 | - | 2 | 257 | 3349257 | ||
BC107739.1 | - | 2 | 265 | |||
BE903613.1 | - | 2 | 279 | |||
BM452267.1 | - | 542 | ||||
CD014005.1 | - | 2 | 265 | |||
DC331440.1 | - | 2 | 369 | |||
J03934.1 | - | 2 | 190 |