Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001931.4 | NP_001922.2 | 1 | 801 | ||
XM_011542647.2 | XP_011540949.1 | 1 | 448 | |||
GenBank mRNA | AF317200.1 | - | 1 | 190 | ||
AK057299.1 | - | 1 | 801 | |||
AK223596.1 | - | 1 | 190 | |||
AK297103.1 | - | 1 | 204 | |||
AK299562.1 | - | 1 | 19 | |||
AK303930.1 | - | 1 | 207 | |||
AL832703.1 | - | 1 | 173 | |||
BC039084.1 | - | 1 | 170 | 4508489 | ||
Y00978.1 | - | 1 | 46 |