Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001931.4 | NP_001922.2 | 13 | 2451 | ||
XM_011542647.2 | XP_011540949.1 | 12 | 1990 | |||
GenBank mRNA | AK057299.1 | - | 13 | 2451 | ||
AK223596.1 | - | 13 | 1840 | |||
AK296274.1 | - | 13 | 1730 | |||
AK297103.1 | - | 10 | 1197 | |||
AL832703.1 | - | 13 | 1822 | |||
BC039084.1 | - | 13 | 1820 | 4508489 | ||
Y00978.1 | - | 13 | 1696 |