Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000108.4 | NP_000099.2 | 12 | 1418 | ||
NM_001289750.1 | NP_001276679.1 | 10 | 1269 | |||
NM_001289751.1 | NP_001276680.1 | 11 | 1349 | |||
NM_001289752.1 | NP_001276681.1 | 11 | 1274 | |||
GenBank mRNA | AB209703.1 | - | 12 | 1328 | ||
AK225289.1 | - | 12 | 1342 | |||
AK225301.1 | - | 12 | 1354 | |||
AK294146.1 | - | 13 | 1355 | |||
AK295080.1 | - | 10 | 1212 | |||
AK297508.1 | - | 11 | 1283 | |||
AK300077.1 | - | 11 | 1207 | |||
AK312346.1 | - | 12 | 1365 | |||
BC018648.2 | - | 12 | 1327 | 3029752 | ||
BC018696.2 | - | 12 | 1327 | 3029752 | ||
J03490.1 | - | 12 | 1350 | |||
J03620.1 | - | 12 | 1324 |