Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DLG1 SIRNA Assays ›
Gene Symbol:
DLG1
Gene Name:
discs large MAGUK scaffold protein 1
Gene Aliases:
DLGH1, SAP-97, SAP97, dJ1061C18.1.1, hdlg
Chromosome Location:
Chr.3: 197042560 - 197299272 on Build GRCh38
UniGene ID:
Hs.292549
Species:
Human
siRNA ID
HSS102791
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DLG1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098424.1
NP_001091894.1
22
2344
NM_001204386.1
NP_001191315.1
21
2308
NM_001204387.1
NP_001191316.1
18
2165
NM_001204388.1
NP_001191317.1
17
2129
NM_001290983.1
NP_001277912.1
22
2413
NM_004087.2
NP_004078.2
22
2410
XM_005269289.3
XP_005269346.1
22
2692
XM_011512502.2
XP_011510804.1
22
2626
XM_011512503.1
XP_011510805.1
21
2587
XM_011512505.1
XP_011510807.1
21
2524
XM_011512506.1
XP_011510808.1
20
2470
XM_017005800.1
XP_016861289.1
22
2827
XM_017005801.1
XP_016861290.1
22
2347
XM_017005802.1
XP_016861291.1
22
2417
XM_017005803.1
XP_016861292.1
22
2338
XM_017005804.1
XP_016861293.1
22
2689
XM_017005805.1
XP_016861294.1
22
2443
XM_017005806.1
XP_016861295.1
21
2590
XM_017005807.1
XP_016861296.1
21
2689
XM_017005808.1
XP_016861297.1
21
2248
XM_017005809.1
XP_016861298.1
21
2350
XM_017005810.1
XP_016861299.1
21
2245
XM_017005811.1
XP_016861300.1
20
2536
XM_017005812.1
XP_016861301.1
20
2190
XM_017005813.1
XP_016861302.1
20
2194
XM_017005814.1
XP_016861303.1
20
2297
XM_017005815.1
XP_016861304.1
20
2533
XM_017005816.1
XP_016861305.1
21
2623
XM_017005817.1
XP_016861306.1
21
2154
XM_017005818.1
XP_016861307.1
21
2256
XM_017005819.1
XP_016861308.1
21
2173
XM_017005820.1
XP_016861309.1
20
2188
XM_017005821.1
XP_016861310.1
18
4695
XM_017005822.1
XP_016861311.1
18
4631
XM_017005823.1
XP_016861312.1
17
4592
XM_017005824.1
XP_016861313.1
12
1211
GenBank mRNA
AB209536.2
-
15
1503
AK293967.1
-
17
1848
AK294772.1
-
18
1892
AK294855.1
-
17
2129
AL831922.1
-
4
1905
BC140841.1
-
22
2375
9021709
BC144651.1
-
21
2273
9053182
EF553524.1
-
22
2413
U13896.1
-
22
2408
U13897.1
-
22
2342
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
branching involved in ureteric bud morphogenesis
immunological synapse formation
endothelial cell proliferation
lens development in camera-type eye
T cell cytokine production
protein dephosphorylation
actin filament organization
mitotic cell cycle checkpoint
establishment or maintenance of cell polarity
chemical synaptic transmission
nervous system development
positive regulation of cell proliferation
regulation of cell shape
viral process
single organismal cell-cell adhesion
peristalsis
positive regulation of actin filament polymerization
cortical actin cytoskeleton organization
astral microtubule organization
membrane raft organization
regulation of myelination
activation of protein kinase activity
ion transmembrane transport
cellular protein complex localization
T cell activation
negative regulation of T cell proliferation
regulation of membrane potential
amyloid precursor protein metabolic process
receptor clustering
positive regulation of potassium ion transport
cortical microtubule organization
establishment or maintenance of epithelial cell apical/basal polarity
negative regulation of mitotic cell cycle
GMP metabolic process
GDP metabolic process
reproductive structure development
embryonic skeletal system morphogenesis
smooth muscle tissue development
negative regulation of epithelial cell proliferation
establishment of centrosome localization
negative regulation of protein kinase B signaling
hard palate development
negative regulation of ERK1 and ERK2 cascade
bicellular tight junction assembly
protein localization to plasma membrane
positive regulation of establishment of protein localization to plasma membrane
receptor localization to synapse
regulation of NIK/NF-kappaB signaling
regulation of sodium ion transmembrane transport
negative regulation of p38MAPK cascade
guanylate kinase activity
phosphoprotein phosphatase activity
protein binding
protein C-terminus binding
cytoskeletal protein binding
ligand-gated ion channel activity
potassium channel regulator activity
protein kinase binding
phosphatase binding
mitogen-activated protein kinase kinase binding
protein complex scaffold
ionotropic glutamate receptor binding
ion channel binding
L27 domain binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbcvc:80/10.239.164.135:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline