Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166278.1 | NP_001159750.1 | 8 | 798 | ||
NM_020730.2 | NP_065781.1 | 10 | 1341 | |||
NM_021120.3 | NP_066943.2 | 15 | 2286 | |||
XM_005262248.3 | XP_005262305.1 | 10 | 938 | |||
XM_006724625.2 | XP_006724688.1 | 16 | 2387 | |||
XM_006724626.2 | XP_006724689.1 | 16 | 2333 | |||
XM_011530883.1 | XP_011529185.1 | 15 | 2345 | |||
XM_017029322.1 | XP_016884811.1 | 9 | 1288 | |||
XM_017029323.1 | XP_016884812.1 | 10 | 1276 | |||
XM_017029324.1 | XP_016884813.1 | 9 | 1254 | |||
XM_017029325.1 | XP_016884814.1 | 8 | 2648 | |||
XM_017029326.1 | XP_016884815.1 | 8 | 727 | |||
XM_017029327.1 | XP_016884816.1 | 7 | 697 | |||
GenBank mRNA | AB033058.1 | - | 10 | 1339 | ||
AB209328.1 | - | 15 | 2286 | |||
AK303377.1 | - | 8 | 798 | |||
AK304020.1 | - | 10 | 1054 | |||
AK315041.1 | - | 15 | 2225 | |||
AK316518.1 | - | 10 | 1040 | |||
BC093864.1 | - | 15 | 1987 | 7939709 | ||
BC093866.1 | - | 15 | 1987 | 7939711 | ||
U49089.1 | - | 15 | 2172 |