Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128827.2 | NP_001122299.1 | 4 | 666 | ||
NM_001321074.1 | NP_001308003.1 | 6 | 1514 | |||
NM_001321075.1 | NP_001308004.1 | 4 | 675 | |||
NM_001321076.1 | NP_001308005.1 | 3 | 320 | |||
NM_001321077.1 | NP_001308006.1 | 3 | 311 | |||
NM_001365.4 | NP_001356.1 | 6 | 1523 | |||
NR_135527.1 | 6 | 1523 | ||||
XM_005256491.1 | XP_005256548.1 | 4 | 765 | |||
XM_011523698.1 | XP_011522000.1 | 6 | 1411 | |||
XM_011523699.2 | XP_011522001.1 | 7 | 653 | |||
XM_011523702.1 | XP_011522004.1 | 3 | 190 | |||
XM_017024288.1 | XP_016879777.1 | 3 | 411 | |||
XM_017024289.1 | XP_016879778.1 | 3 | 402 | |||
XM_017024290.1 | XP_016879779.1 | 3 | 880 | |||
GenBank mRNA | AF028825.1 | - | 4 | 225 | ||
AK293835.1 | - | 4 | 672 | |||
AK296357.1 | - | 6 | 1211 | |||
AK297346.1 | - | 3 | 268 | |||
AK299801.1 | - | 3 | 282 | |||
AK307804.1 | - | 4 | 649 | |||
AK308533.1 | - | 3 | 425 | |||
AK316096.1 | - | 4 | 228 | |||
BC136553.1 | - | 6 | 1047 | 9020543 | ||
BP213421.1 | - | 3 | 311 | |||
U68138.1 | - | 2 | 82 | |||
U83192.1 | - | 6 | 1181 |