Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081560.2 | NP_001075029.1 | 8 | 1141 | ||
NM_001081562.2 | NP_001075031.1 | 8 | 1141 | |||
NM_001081563.2 | NP_001075032.1 | 7 | 1510 | |||
NM_001288764.1 | NP_001275693.1 | 9 | 1200 | |||
NM_001288765.1 | NP_001275694.1 | 6 | 781 | |||
NM_001288766.1 | NP_001275695.1 | 8 | 1141 | |||
NM_004409.4 | NP_004400.4 | 8 | 1141 | |||
GenBank mRNA | AB209363.1 | - | 8 | 1984 | ||
AK297299.1 | - | 6 | 732 | |||
AK302874.1 | - | 8 | 1074 | |||
BC026328.1 | - | 9 | 1195 | 4792104 | ||
BC062553.1 | - | 8 | 1062 | 6480902 | ||
KJ534826.1 | - | 7 | 966 | |||
KJ534827.1 | - | 7 | 966 | |||
KJ534828.1 | - | 7 | 966 | |||
L19268.1 | - | 8 | 1063 | |||
M87312.1 | - | 7 | 1511 | |||
M94203.1 | - | 8 | 813 | |||
S72883.1 | - | 8 | 972 |