Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001369.2 | NP_001360.1 | 77 | 13414 | ||
XM_005248262.3 | XP_005248319.2 | 77 | 13497 | |||
XM_017009177.1 | XP_016864666.1 | 75 | 13077 | |||
XM_017009178.1 | XP_016864667.1 | 77 | 14300 | |||
XM_017009179.1 | XP_016864668.1 | 71 | 12594 | |||
XM_017009185.1 | XP_016864674.1 | 49 | 8793 | |||
XM_017009186.1 | XP_016864675.1 | 46 | 8209 | |||
XM_017009188.1 | XP_016864677.1 | 43 | 9025 | |||
GenBank mRNA | AB046823.2 | - | 25 | 4478 | ||
AB290164.1 | - | 30 | 5417 | |||
AK026756.1 | - | 5 | 770 | |||
AK128600.1 | - | 19 | 3417 | |||
AY045575.1 | - | 77 | 13414 |