Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001369.2 | NP_001360.1 | 78 | 13686 | ||
XM_005248262.3 | XP_005248319.2 | 78 | 13769 | |||
XM_017009177.1 | XP_016864666.1 | 76 | 13349 | |||
XM_017009178.1 | XP_016864667.1 | 78 | 14572 | |||
XM_017009179.1 | XP_016864668.1 | 72 | 12866 | |||
XM_017009185.1 | XP_016864674.1 | 50 | 9065 | |||
XM_017009186.1 | XP_016864675.1 | 47 | 8481 | |||
XM_017009188.1 | XP_016864677.1 | 44 | 9297 | |||
GenBank mRNA | AB046823.2 | - | 26 | 4750 | ||
AB290164.1 | - | 31 | 5689 | |||
AK026756.1 | - | 6 | 1042 | |||
AK128600.1 | - | 20 | 3689 | |||
AY045575.1 | - | 78 | 13686 |