Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNASE1L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256560.1 | NP_001243489.1 | 2 | 259 | ||
NM_004944.3 | NP_004935.1 | 2 | 259 | |||
GenBank mRNA | AF047354.1 | - | 2 | 232 | ||
AK131323.1 | - | 5 | 754 | |||
AK301263.1 | - | 2 | 225 | |||
AK313303.1 | - | 2 | 225 | |||
BC015831.1 | - | 2 | 229 | 4723725 | ||
BI521582.1 | - | 2 | 469 | |||
BI834035.1 | - | 4 | 647 | |||
U56814.1 | - | 2 | 186 |