Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNASE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001375.2 | NP_001366.1 | 6 | 968 | ||
GenBank mRNA | AB004574.1 | - | 6 | 894 | ||
AF045937.1 | - | 6 | 898 | |||
AF047016.1 | - | 6 | 894 | |||
AF060222.1 | - | 6 | 915 | |||
AK075449.1 | - | 6 | 883 | |||
AK297493.1 | - | 5 | 731 | |||
AK311145.1 | - | 6 | 892 | |||
AK315358.1 | - | 6 | 910 | |||
BC010419.2 | - | 6 | 846 | 3528108 | ||
BC065209.1 | - | 6 | 913 | 5502061 | ||
BT007047.1 | - | 6 | 822 | |||
FJ752529.1 | - | 6 | 774 |