Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNC1LI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323955.1 | NP_001310884.1 | 3 | 509 | ||
NM_006141.2 | NP_006132.1 | 4 | 420 | |||
NR_136658.1 | 4 | 626 | ||||
XM_017023008.1 | XP_016878497.1 | 4 | 419 | |||
GenBank mRNA | AF035812.1 | - | 4 | 395 | ||
AK291766.1 | - | 4 | 399 | |||
AK295049.1 | - | 4 | 419 | |||
AK304233.1 | - | 3 | 275 | |||
AL708642.1 | - | 4 | 420 | |||
BC025959.1 | - | 4 | 395 | 4329417 | ||
BX648863.1 | - | 4 | 402 | |||
DC417195.1 | - | 3 | 275 |