Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNMT3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207055.1 | NP_001193984.1 | 2 | 333 | ||
NM_001207056.1 | NP_001193985.1 | 2 | 333 | |||
NM_006892.3 | NP_008823.1 | 2 | 333 | |||
NM_175848.1 | NP_787044.1 | 2 | 333 | |||
NM_175849.1 | NP_787045.1 | 2 | 333 | |||
NM_175850.2 | NP_787046.1 | 2 | 312 | |||
XM_011528653.2 | XP_011526955.1 | 2 | 185 | |||
XM_011528654.2 | XP_011526956.1 | 2 | 186 | |||
GenBank mRNA | AB208880.1 | - | 2 | 311 | ||
AF156487.1 | - | 2 | 126 | |||
AF156488.1 | - | 2 | 126 | |||
AF176228.1 | - | 2 | 312 | |||
AF331857.1 | - | 2 | 333 | |||
AK295176.1 | - | 2 | 100 | |||
AK299776.1 | - | 2 | 72 | |||
AK299821.1 | - | 2 | 76 | |||
AK299915.1 | - | 2 | 110 | |||
AK300857.1 | - | 2 | 122 | |||
DQ321787.1 | - | 2 | 333 |