Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290223.1 | NP_001277152.1 | 49, 50 | 5357 | ||
NM_001380.4 | NP_001371.1 | 49, 50 | 5294 | |||
XM_011539422.2 | XP_011537724.1 | 49, 50 | 5335 | |||
XM_011539424.1 | XP_011537726.1 | 49, 50 | 5245 | |||
XM_017015813.1 | XP_016871302.1 | 49, 50 | 5267 | |||
XM_017015816.1 | XP_016871305.1 | 32, 33 | 3515 | |||
GenBank mRNA | BC146857.1 | - | 49, 50 | 5247 | 9056785 | |
BU556771.1 | - | 6, 7 | 810 | |||
D50857.1 | - | 49, 50 | 5208 |