Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DXO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005510.3 | NP_005501.2 | 3 | 888 | ||
XM_006715005.3 | XP_006715068.1 | 3 | 967 | |||
XM_017010329.1 | XP_016865818.1 | 3 | 671 | |||
GenBank mRNA | AF059252.1 | - | 3 | 684 | ||
AK310107.1 | - | 2 | 967 | |||
BC009344.2 | - | 3 | 728 | 4130758 | ||
BC019083.2 | - | 3 | 900 | 4647012 |